missing translation for 'onlineSavingsMsg'
Learn More

KIR2DL1/CD158a Antibody (2F9), DyLight 350, Novus Biologicals™

Product Code. 18410616 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18410616 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18410616 Supplier Novus Biologicals Supplier No. NBP104315UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

KIR2DL1/CD158a Monoclonal antibody specifically detects KIR2DL1/CD158a in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIR2DL1/CD158a
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone 2F9
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias CD158 antigen-like family member A, CD158a antigen, CD158Ankat1, cl-42, killer cell immunoglobulin-like receptor 2DL1, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 1, killer Ig receptor, killer inhibitory receptor 2-2-1, KIR221, MHC class I NK cell receptor, Natural killer-associated transcript 1, NKAT, NKAT-1, NKAT1KIR-K64, p58 killer cell inhibitory receptor KIR-K64, p58 natural killer cell receptor clones CL-42/47.11, p58 NK cell inhibitory receptor NKR-K6,47.11, p58 NK receptor CL-42/47.11, p58.1, p58.1 MHC class-I-specific NK receptor
Host Species Mouse
Immunogen Recombinant human KIR2DL1/CD158a (23-223 aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3802
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.