missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kir2.2 (KCNJ12) Mouse anti-Human, Mouse, Rat, Clone: S124B-38, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA527678
This item is not returnable.
View return policy
Description
Inward rectifying potassium channel that is activated by phosphatidylinositol 4,5-bisphosphate and that probably participates in controlling the resting membrane potential in electrically excitable cells. Probably participates in establishing action potential waveform and excitability of neuronal and muscle tissues. Inward rectifier potassium channels are characterized by a greater tendency to allow potassium to flow into the cell rather than out of it. Their voltage dependence is regulated by the concentration of extracellular potassium; as external potassium is raised, the voltage range of the channel opening shifts to more positive voltages. The inward rectification is mainly due to the blockage of outward current by internal magnesium.
Specifications
| Kir2.2 (KCNJ12) | |
| Monoclonal | |
| 1 mg/ml | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.4 | |
| P52187, P52188, Q14500 | |
| KCNJ12 | |
| Fusion protein amino acids 362-427 of rat Kir2.2 | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| S124B-38 | |
| Unconjugated | |
| KCNJ12 | |
| IRK2, KCNJ12, kcnj12x, KCNJN1, Kir2.2v, potassium inwardly rectifying channel subfamily J member 12 | |
| Mouse | |
| Protein G | |
| RUO | |
| 117052, 16515, 3768 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction