missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The antibody recognizes the C-term of KCNJ2.
The KIR2 subunit family includes 2.1, 2.2, 2.3 and 2.4. Unlike G protein-coupled KIR3 subunits, KIR2.1 requires both phosphorylation by PKA and ATP hydrolysis for functional activity. KIR2.1 is expressed in the superior and inferior collicula and the pontine region of the brain, where it moderates synaptic transmission, like many other potassium channels. In the placenta, KIR2.1 is expressed throughout gestation in cytotrophoblast cells. In the kidney, KIR2.1 colocalizes with KIR5.1 in the proximal tubule. KIR2.1, 2.2 and 2.3 associate with the membrane-associated guanylate kinase synapseassociated protein 97 in the cerebellum and heart. Phosphorylation of KIR2.2 by protein kinase A inhibits the associates with SAP97.
Specifications
Specifications
| Antigen | Kir2.1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.33 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | KCNJ2 |
| Gene Accession No. | P35561, P63252, Q64273 |
| Gene Alias | HHBIRK1, HHIRK1, hIRK1, IRK 1, IRK1, KCNJ2, Kir2.1, LQT7, SQT3 |
| Gene Symbols | Kcnj2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?