missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Kir2.1 (KCNJ2) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588883
This item is not returnable.
View return policy
Description
Immunogen sequence: NEILWGHRYE PVLFEEKHYY KVDYSRFHKT YEVPNTPLCS ARDLAEKKYI LSNANSFCYE NEVALTSKEE DDSENGVPES TSTDTPPDID LHNQASVPLE PRPLRRESEI; Positive Samples: A-549, Mouse heart, Rat heart; Cellular Location: Lipid-anchor, Membrane, Multi-pass membrane protein.
The KIR2 subunit family includes 2. 1, 2. 2, 2. 3 and 2. 4. Unlike G protein-coupled KIR3 subunits, KIR2. 1 requires both phosphorylation by PKA and ATP hydrolysis for functional activity. KIR2. 1 is expressed in the superior and inferior collicula and the pontine region of the brain, where it moderates synaptic transmission, like many other potassium channels. In the placenta, KIR2. 1 is expressed throughout gestation in cytotrophoblast cells. In the kidney, KIR2. 1 colocalizes with KIR5. 1 in the proximal tubule. KIR2. 1, 2. 2 and 2. 3 associate with the membrane-associated guanylate kinase synapseassociated protein 97 in the cerebellum and heart. Phosphorylation of KIR2. 2 by protein kinase A inhibits the associates with SAP97.
Specifications
| Kir2.1 (KCNJ2) | |
| Polyclonal | |
| Unconjugated | |
| Kcnj2 | |
| ATFB9; cardiac inward rectifier KIR2.1; cardiac inward rectifier potassium channel; EGK_08925; HHBIRK1; HHIRK 1; HHIRK1; HIRK 1; hIRK1; Inward rectifier K; inward rectifier K(+) channel Kir2.1; inward rectifier K+ channel KIR2.1; inward rectifier potassium channel; inward rectifier potassium channel 2; inwardly rectifying potassium channel Kir2.1; IRK 1; IRK1; IRK-1; IRK2; Kcnf1; KCNJ2; KIR2.1; LQT 7; LQT7; Potassium channel, inwardly rectifying subfamily J member 2; potassium channel, inwardly rectifying subfamily J, member 2; potassium inwardly rectifying channel subfamily J member 2; potassium inwardly-rectifying channel, subfamily J, member 2; potassium voltage-gated channel subfamily J member 2; RBL-IRK1; SQT 3; SQT3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 16518, 29712, 3759 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1.72 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P35561, P63252, Q64273 | |
| Kcnj2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 318-427 of human KCNJ2 (NP_0008821). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction