missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kir2.1 Antibody (S112), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12900AF532
This item is not returnable.
View return policy
Description
Kir2.1 Monoclonal antibody specifically detects Kir2.1 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Kir2.1 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| S112 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| Cardiac inward rectifier potassium channel, HHBIRK1, HHIRK1, hIRK1, Inward rectifier K(+) channel Kir2.1, inward rectifier K+ channel KIR2.1, inward rectifier potassium channel 2, IRK-1, IRK1LQT7, Kir2.1, Potassium channel, inwardly rectifying subfamily J member 2, potassium inwardly-rectifying channel, subfamily J, member 2, SQT3 | |
| Fusion protein amino acids 41-64 and 189-428 of mouse Kir2.1 | |
| 0.1 mL | |
| Neuroscience | |
| 3759 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction