missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIR/CD158 Antibody (lirilumab), Alexa Fluor™ 350, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-28035AF350
This item is not returnable.
View return policy
Description
KIR/CD158 Monoclonal antibody specifically detects KIR/CD158 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| KIR/CD158 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| CD158 antigen-like family member A, CD158a antigen, CD158Ankat1, cl-42, killer cell immunoglobulin-like receptor 2DL1, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 1, killer Ig receptor, killer inhibitory receptor 2-2-1, KIR221, MHC class I NK cell receptor, Natural killer-associated transcript 1, NKAT, NKAT-1, NKAT1KIR-K64, p58 killer cell inhibitory receptor KIR-K64, p58 natural killer cell receptor clones CL-42/47.11, p58 NK cell inhibitory receptor NKR-K6,47.11, p58 NK receptor CL-42/47.11, p58.1, p58.1 MHC class-I-specific NK receptor | |
| KIR | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| lirilumab | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| 3802 | |
| Store at 4°C in the dark. | |
| IgG4 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction