missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KIP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104486
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CIB2.
DNA-dependent protein kinases (DNA-PK) play a role in the repair of double-strand DNA breaks and in the process of V(D)J recombination during lymphoid development. By EST database searching for sequences homologous to the DNA-PK gene KIP/CIB and 5-prime RACE, Seki, N et al. isolated a full-length cDNA, which they designated as KIP2, from a human fetal brain cDNA library. KIP2 encodes a deduced 187-amino acid protein with a predicted molecular mass of 22 kD. The KIP2 protein shares 46%, 39%, and 30% sequence identity with the calcium-binding proteins KIP/CIB, calcineurin B and calmodulin respectively. KIP2 contains two EF-hand motifs and a helix-loop-helix motif involved in coordinating the calcium ion, indicating that KIP2 may also bind calcium. KIP2 is ubiquitously expressed in various human tissues.
Specifications
| KIP2 | |
| Polyclonal | |
| Unconjugated | |
| CIB2 | |
| 2810434I23Rik; AI449053; calcium and integrin binding family member 2; calcium and integrin-binding family member 2; calcium binding protein Kip 2; calcium binding protein Kip2; CIB2; DFNB48; DNA-dependent protein kinase catalytic subunit-interacting protein 2; kinase interacting protein 2; kinase-interacting protein 2; KIP 2; Kip2; USH1J | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10518, 300719, 56506 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75838, Q568Z7, Q9Z309 | |
| CIB2 | |
| A synthesized peptide derived from human CIB2(Accession O75838), corresponding to amino acid residues N40-D90. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction