missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Kininogen 1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA581957

Product Code. 16358924

  • £478.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: ALKKYNSQNQ SNNQFVLYRI TEATKTVGSD TFYSFKYEIK EGDCPVQSGK TWQDCEYKDA AKAATGECTA TVGKRSSTKF SVATQTCQIT PAEGPVVTAQ YDCLGCVHPI STQSPDLEPI LRHGIQYFNN N Highest antigen sequence indentity to the following orthologs: Mouse - 62%, Rat - 61%.

This gene uses alternative splicing to generate two different proteins- high molecular weight kininogen (HMWK) and low molecular weight kininogen (LMWK). HMWK is essential for blood coagulation and assembly of the kallikrein-kinin system. Also, bradykinin, a peptide causing numerous physiological effects, is released from HMWK. In contrast to HMWK, LMWK is not involved in blood coagulation. Three transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Kininogen 1
Polyclonal
Unconjugated
KNG1
alpha-1-MAP; Alpha-2-thiol proteinase inhibitor; BDK; BK; Bradykinin; fitzgerald factor; high molecular weight kininogen; H-kininigen; HMWK; Ile-Ser-Bradykinin; Kallidin I; Kallidin II; kininogen; kininogen 1; kininogen 1-like 1; kininogen-1; Kininogen-1 heavy chain; Kininogen-1 light chain; Kng; Kng1; Kng1l1; L-kininogen; low molecular weight (LMW) T-kininogen II; Low molecular weight growth-promoting factor; Lysyl-bradykinin; Major acute phase protein; major acute phase protein (alpha-1-MAP); Thiostatin; T-kinin; T-kininogen 2; T-kininogen 2 heavy chain; T-kininogen 2 light chain; T-kininogen II; T-kininogen II heavy chain; T-kininogen II light chain; williams-Fitzgerald-Flaujeac factor
Rabbit
Antigen affinity chromatography
RUO
3827
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P01042
KNG1
Recombinant protein corresponding to Human KNG1. Recombinant protein control fragment (Product #RP-100525).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.