missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kininogen 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99364-50ul
This item is not returnable.
View return policy
Description
Kininogen 1 Polyclonal antibody specifically detects Kininogen 1 in Human samples. It is validated for ELISA
Specifications
| Kininogen 1 | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| Alpha-2-thiol proteinase inhibitor, BDK, BK, bradykinin, Fitzgerald factor, High molecular weight kininogen, HMWK, kininogen-1, Williams-Fitzgerald-Flaujeac factor | |
| Produced in rabbits immunized with purified, recombinant Human Kininogen 1 (Accession#: NP_001095886.1; Gln 19-Ser 644) | |
| 50 μg | |
| Apoptosis, Immunology | |
| 3827 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction