Learn More
Thermo Scientific™ KingFisher Plastic Consumables for SARS-CoV-2 Testing
Shop All Thermo Scientific ProductsDescription
These KingFisher compatible plastic consumables are offered specifically for labs performing COVID-19 testing with the MagMAX Viral/Pathogen Nucleic Acid Purification Kit and the TaqPath COVID-19 Combo Kit.
For TaqPath COVID-19 testing with MagMAX Viral/Pathogen Nucleic Acid Isolation Kit
Specifications
Specifications
| Format | 96 Deep-well Plate |
| Capacity | 96 Samples |
| For Use With (Equipment) | Duo Prime, Flex, Apex, Presto |
| Includes | KF 96 DW plate, v-bottom |
| Quantity | 50/Pk. |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.