Learn More
Description
The KingFisher™ mL Purification System is discontinued. Thermo Fisher Scientific offers other automation platforms such as the Thermo Scientific™ KingFisher™ Duo Prime Purification System, KingFisher™ Flex Purification System and KingFisher Apex. The company will continue to support current KingFisher™ mL users with plastics, service, and technical support through 2028, or best effort.
Thermo Fisher Scientific has transitioned to limited support stage. It will offer limited support contracts and provide best-effort service and support, subject to availability of spare parts and resources.
If you are interested in upgrading to one of its newer sample preparation instruments, please contact your local account manager for product information.
- Offers rapid and reproducible purification of high-quality DNA, RNA, proteins and cells for various types of downstream applications.
- Open and flexible system lets customer use any magnetic particle-based kit to meet application demands
- Easy-to-use Thermo Scientific™ BindIt™ Software provides instrument control, protocol creation and modification
- Ready-made protocols for different type of applications available
Recommended for:
Genomics and proteomics; Drug discovery; Forensics; Biomarker discovery; Quality control; Veterinary assays
Specifications
Specifications
| Volume (Metric) Processing | 50 to 1000 μL |
| Capacity | 15 Samples/Run |
| Description | KingFisher mL Purification System, 100-240V, 50/60Hz nominal |
| Collection Efficiency of Particles | 95% |
| Display | START/STOP/TWO CURSOR KEYS/LCD |
| For Use With (Application) | Genomics and proteomics; Drug discovery; Forensics; Biomarker discovery; Quality control; Veterinary assays |
| Magnetic Rods | 3 x 5 format |
| Particle Size (Metric) | ca. >1 μm |
| Quantity | Each |
| Tip Combo | Special design, 1 x 5 format |
| Show More |
For research use only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
