missing translation for 'onlineSavingsMsg'
Learn More

Kinetin, TRC

Product Code. 30137931
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137931 1 g 1g
17901855 100 mg 100mg
17973275 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137931 Supplier Toronto Research Chemicals Supplier No. TRCK5700051G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 525-79-1
Molecular Formula C10 H9 N5 O
Molecular Weight (g/mol) 215.21
Synonym 6-Furfurylaminopurine, 9H-Purin-6-amine, N-(2-furanylmethyl)- (ACI), 1H-Purin-6-amine, N-(2-furanylmethyl)- (9CI), Adenine, N-furfuryl- (7CI, 8CI), Adenine, N6-furfuryl- (6CI), N-(2-Furanylmethyl)-9H-purin-6-amine (ACI), 2-Furanmethanamine, N-1H-purin-6-yl-, 6-(Furfurylamino)purine, 6-Furfuryladenine, FAP, Kinetin, Kinetin (plant hormone), KT, MB 905, MeSH ID: D007701, N-Furfuryladenine, N6-(Furfurylamino)purine, N6-Furfuryladenine, NSC 23119
IUPAC Name N-(furan-2-ylmethyl)-9H-purin-6-amine
SMILES C(Nc1ncnc2[nH]cnc12)c3occc3

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H9N5O/c1-2-7(16-3-1)4-11-9-8-10(13-5-12-8)15-6-14-9/h1-3,5-6H,4H2,(H2,11,12,13,14,15)
Formula Weight 215.0807
Percent Purity >95%
Chemical Name or Material Kinetin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.