missing translation for 'onlineSavingsMsg'
Learn More

Kinetin, MedChemExpress

Product Code. 16411328
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16411328 5 g 5g
16442509 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16411328 Supplier Medchem Express Supplier No. HYN0160
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Kinetin (N6-furfuryladenine) belongs to the family of N6-substituted adenine derivatives known as cytokinins, which are plant hormones involved in cell division, differentiation and other physiological processes. Kinetin has anti-aging effects.

Kinetin (N6-furfuryladenine) belongs to the family of N6-substituted adenine derivatives known as cytokinins, which are plant hormones involved in cell division, differentiation and other physiological processes. Kinetin has anti-aging effects.

Specifications

Chemical Name or Material Kinetin
CAS 525-79-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C10H9N5O
Quantity 5 g
Synonym 6-Furfuryladenine N6-Furfuryladenine
Solubility Information DMSO : 33.33 mg/mL (154.87 mM; Need ultrasonic) ∣H2O : 1 mg/mL (4.65 mM; ultrasonic and warming and heat to 80°C)
SMILES C12=NC=NC(NCC3=CC=CO3)=C1N=CN2
Molecular Weight (g/mol) 215.21
Formula Weight 215.21
Percent Purity 98.92%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.