missing translation for 'onlineSavingsMsg'
Learn More

Kinetin, 99%

Product Code. 11484724
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11484724 25 g 25g
11464724 1 g 1g
11474724 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11484724 Supplier Thermo Scientific Alfa Aesar Supplier No. A13720.14
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 525-79-1 | C10H9N5O | 215.216 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Kinetin acts as plant growth accelerator, auxin, plant growth regulator, plant cell division promotor. It also acts as cell division factor found in various plant parts and in yeast. A plant growth regulator. Augments growth of microbial cultures.

Solubility
Soluble in water (<1mg/mL) at 25°C, Acetic acid (49.00-51.00 mg/ml), DMSO (43 mg/ml) at 25°C, ethanol (<1mg/mL) at 25°C, 0.1 N Sodium hydroxide (1% w/v), methanol (slightly), and dilute aqueous hydrochloric acid or sodium hydroxide (freely).

Notes
Protect from heat. Store at -20°C. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 525-79-1
Molecular Formula C10H9N5O
Molecular Weight (g/mol) 215.216
MDL Number MFCD00075757
InChI Key QANMHLXAZMSUEX-UHFFFAOYSA-N
Synonym kinetin, 6-furfurylaminopurine, 6-furfurylamino purine, 6-furfuryladenine, n6-furfuryladenine, n-furfuryladenine, cytokinin, n-furan-2-ylmethyl-9h-purin-6-amine, n6-furfurylamino purine, adenine, n-furfuryl
PubChem CID 3830
ChEBI CHEBI:27407
IUPAC Name N-(furan-2-ylmethyl)-7H-purin-6-amine
SMILES C1=COC(=C1)CNC2=NC=NC3=C2NC=N3

Specifications

Melting Point ∼270°C (decomposition)
Assay Percent Range 99%
Odor Odorless
Quantity 25 g
Beilstein 21703
Merck Index 14,5311
Solubility Information Soluble in water (<1mg/ml) at 25°C,Acetic acid (49.00-51.00mg/ml),DMSO (43mg/ml) at 25°C,ethanol (<1mg/ml) at 25°C,0.1 N Sodium hydroxide (1% w/v),methanol (slightly),and dilute aqueous hydrochloric acid or sodium hydroxide (freely).
Formula Weight 215.22
Percent Purity 99%
Chemical Name or Material Kinetin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.