missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ Kinetic-QCL™ Chromogenic LAL Assay Kit

Product Code. 11600111
Change view
Click to view available options
Quantity:
192 Tests
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11600111 192 Tests 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11600111 Supplier Lonza™ Supplier No. 50650U
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Detection of Gram negative bacterial endotoxin. Lonza Kinetic-QCL™ Chromogenic LAL Assay Kit is most sensitive of LAL assays.

  • Sample mixed with reconstituted LAL reagent in 96-well plate and placed in incubating plate reader
  • Absorbance at 405nm
  • Reaction is monitored over time for appearance of yellow color
  • In presence of endotoxin, lysate will cleave chromogenic substrate, causing solution to become yellow
  • Certificate of analysis is provided
TRUSTED_SUSTAINABILITY

Specifications

Quantity 192 Tests
Sensitivity 0.005 EU/mL to 50.0 EU/mL
Content And Storage 2°C to 8°C
Product Type Kinetic-QCL™ Chromogenic LAL Assay Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.