Learn More
Lonza™ Kinetic-QCL Bulk Kinetic Chromogenic LAL Assay
Product Code. 11600251
Change view
Click to view available options
Product Type:
Kinetic-QCL Bulk Kinetic Chromogenic LAL Assay
Unit Size:
Each
Product Code. Product Type Unit Size
11600251 Kinetic-QCL Bulk Kinetic Chromogenic LAL Assay Each
1 options
Product Code. 11600251 Supplier Lonza™ Supplier No. K50643
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
A configuration of the KQCL™ Kinetic Chromogenic LAL Endotoxin Detection Assay.
This bulk kit configuration does not contain endotoxin and requires that E50-643L is ordered separately. Testing of specific lots for making larger product reserves is possible. Please contact your local sales representative for further details. The Kinetic-QCL™ Kinetic Chromogenic LAL Assay is a quantitative, kinetic assay for the detection of Gram-negative bacterial endotoxin. A sample is mixed with the reconstituted LAL reagent in a 96-well plate and placed in an incubating plate reader that measures absorbance at 405 nm. The reaction is automatically monitored over time for the appearance of a yellow color.
In the presence of endotoxin, the lysate will begin to cleave the chromogenic substrate, causing the solution to become yellow. The time required for the change is inversely proportional to the amount of endotoxin present. The concentration of unknown samples can then be calculated from a standard curve. It is the chromogenic nature of the Kinetic-QCL™ Kinetic Chromogenic LAL Assay that makes it the most appropriate choice for testing small volume parenterals, vaccines, antibiotics and biologicals. The Kinetic-QCL™ Assay is less impacted by inhibitory products which may interfere with the clotting mechanism in turbidimetric and gel clot assays. With a sensitivity range of 0.005 EU/mL - 50.0 EU/mL, the Kinetic-QCL™ Kinetic Chromogenic LAL Assay is the most sensitive of the LAL assays.
TRUSTED_SUSTAINABILITY
Specifications
Total Assay Time ∼1 hr.
Product Type Kinetic-QCL Bulk Kinetic Chromogenic LAL Assay