missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Kinetensin

Product Code. 16422869
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16422869 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16422869 Supplier MedChemExpress Supplier No. HYP1255

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Kinetensin is a neurotensin-like peptide isolated from pepsin-treated human plasma.

Specifications

Purity 99.21%
CAS 103131-69-7
Content And Storage Protect from light, stored under nitrogen∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light, stored under nitrogen)
For Use With (Application) Neuroscience-Neuromodulation
Molecular Weight (g/mol) 1172.38 g/mol
Product Type Natural Products
Quantity 1 mg
Shipping Condition Room temperature
Solubility DMSO : 100 mg/mL (85.30 mM∣Need ultrasonic)
Synonym Kinetensin (human)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.