missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Kinesore
GREENER_CHOICE

Product Code. 18775836
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18775836 10 mg 10mg
18745826 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18775836 Supplier Tocris Bioscience™ Supplier No. 6664/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Kinesin-1 modulator; cell permeable

Kinesore is a kinesin-1 modulator. Inhibits SKIP-KLC2 interaction, but promotes kinesin-1 function in controlling the organization of the microtubule network. Induces remodeling of the microtubule network and the formation of extensive microtubule-rich projections. Cell permeable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,5-Dibromo-N'-((2,5-dimethyl-1-(3-nitrophenyl)-1H-pyrrol-3-yl)methylene)-4-hydroxybenzohydrazide
CAS 363571-83-9
Quantity 10 mg
Molecular Formula C20H16Br2N4O4
Formula Weight 536.17
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.