missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kinesin 5C Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £375.00
Specifications
| Antigen | Kinesin 5C |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18373907
|
Novus Biologicals
NBP3-03752-20UL |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18391417
|
Novus Biologicals
NBP3-03752-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Kinesin 5C Polyclonal specifically detects Kinesin 5C in Human samples. It is validated for Western Blot.Specifications
| Kinesin 5C | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| heavy chain, neuron-specific, KIF 5C, kinesin family member 5C, Kinesin heavy chain neuron-specific 2, MGC111478, NKHC, NKHC2NKHC-2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 510-590 of human Kinesin 5C (NP_004513.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cellular Markers | |
| PBS with 50% glycerol, pH7.3. | |
| 3800 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title