missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kinesin 5B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | Kinesin 5B |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
Kinesin 5B Polyclonal specifically detects Kinesin 5B in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
| Kinesin 5B | |
| Polyclonal | |
| Rabbit | |
| Cell Biology, Cytoskeleton Markers, Neuronal Cell Markers, Neuroscience | |
| Conventional kinesin heavy chain, kinesin family member 5B, kinesin-1 heavy chain, KNS1kinesin 1 (110-120kD), KNSKINH, Ubiquitous kinesin heavy chain, UKHCkinesin heavy chain | |
| KIF5B | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| P33176 | |
| 3799 | |
| Synthetic peptides corresponding to KIF5B (kinesin family member 5B) The peptide sequence was selected from the C terminal of KIF5B. Peptide sequence ANEVKQAVEQQIQSHRETHQKQISSLRDEVEAKAKLITDLQDQNQKMMLE. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title