missing translation for 'onlineSavingsMsg'
Learn More

Kinesin 5B Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18402014 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18402014 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18402014 Supplier Novus Biologicals Supplier No. NB500580JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Kinesin 5B Polyclonal antibody specifically detects Kinesin 5B in Human, Mouse, Porcine samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kinesin 5B
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias Conventional kinesin heavy chain, kinesin family member 5B, kinesin-1 heavy chain, KNS1kinesin 1 (110-120kD), KNSKINH, Ubiquitous kinesin heavy chain, UKHCkinesin heavy chain
Host Species Rabbit
Immunogen Stalk domain of human kinesin (amino acids 331-906) expressed in E. coli (FKHC3).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3799
Target Species Human, Mouse, Pig
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.