missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Kinesin 5A Peptide
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
Specifications
Specifications
| Gene ID (Entrez) | 3798 |
| Species | Human |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB100-1058 |
| Gene Alias | D12S1889, KIF5A variant protein, kinesin family member 5 A, kinesin heavy chain isoform 5 A, Kinesin heavy chain neuron-specific 1, kinesin, heavy chain, neuron-specific, MY050, Neuronal kinesin heavy chain, NKHC1, NKHCspastic paraplegia 10 (autosomal dominant), SPG10 |
| Gene Symbol | KIF5A |
| Quantity | 0.1 mg |
| Research Category | Cell Biology, Cellular Markers |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction