Learn More
Kinematica Polytron™ PT 10-35 GT & PT 3100D Homogenizer: Adapter
Accessory allows old model generators used on old model PT 3100 and PT 10-35 to be used with new models PT 3100D & PT 10-35 GT Homogenizer.
Brand: Kinematica 11035001
Code : 04
Additional Details : Weight : 0.50000kg
Description
Users of older model Polytron™ Homogenizers (3100 & 10-35) can use their existing generators when using new models with the advanced quick connect feature.Specifications
100/115/220/240 V | |
Ideal for dispersing aggregate with A coupling | |
Polytron™ PT 3100/1035 generator | |
1 Ea. |