missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kindlin Mouse anti-Human, Clone: 4A5.14, Novus Biologicals™
Mouse Monoclonal Antibody
£377.00
Specifications
| Antigen | Kindlin |
|---|---|
| Clone | 4A5.14 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1 to 2 μg/mL, Immunoprecipitation 1-2 ug per 100-500 μg of protein 1ml of lysate, Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Description
Kindlin Monoclonal specifically detects Kindlin in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| Kindlin | |
| 0.2 mg/mL | |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| C20orf42fermitin family homolog 1, chromosome 20 open reading frame 42, fermitin family homolog 1 (Drosophila), fermitin family member 1, FLJ20116, KIND1DTGCU2, kindlerin, kindlin 1, Kindlin syndrome protein, Kindlin-1, UNC112 related protein 1, UNC112A, Unc-112-related protein 1, URP1FLJ23423 | |
| Recombinant protein containing Kindlin (residues 216-677) | |
| Primary | |
| Store at 4°C. |
| 4A5.14 | |
| Western Blot 1 to 2 μg/mL, Immunoprecipitation 1-2 ug per 100-500 μg of protein 1ml of lysate, Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 55612 | |
| IgG2a λ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title