missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Kindlin 3 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA519174
This item is not returnable.
View return policy
Description
This antibody is predicted to react with canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 2,000.
The three KINDLINs are a novel family of focal adhesion proteins, localizing to integrin adhesion sites. The KINDLIN proteins are composed of a centrally located FERM domain interrupted by a pleckstrin homology (PH) domain. KINDLIN1 and KINDLIN2 have been shown to play an essential role in integrin-mediated adhesion and spreading. In contrast to the widely expressed KINDLIN1 and KINDLIN2, KINDLIN3 is restricted to hematopoietic cells and is particularly abundant in megakaryocytes and platelets. Several reports describe a transcriptional misregulation of KINDLINs in various types of cancer. A recent study demonstrates that KINDLIN3 is essential for platelet integrin activation and subsequent integrin outside-in signaling, suggesting it may serve as a potential target for the design of therapeutics aimed at specifically disrupting integrin activation in platelets and leukocytes.
Specifications
| Kindlin 3 | |
| Polyclonal | |
| Unconjugated | |
| Fermt3 | |
| C79673; fermitin family homolog 3; fermitin family homolog 3 (Drosophila); fermitin family member 3; Fermt3; Kind3; kindlin 3; Kindlin3; Kindlin-3; MGC10966; MIG-2; MIG2B; MIG2-like protein; RGD1310168; UNC-112 related protein 2; UNC112C; unc-112-related protein 2; URP2; URP2SF | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 83706 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q86UX7 | |
| Fermt3 | |
| Synthetic peptide sequence (ERARGEELDED) corresponding to the C-terminus amino acids of FERMT3 (aa 645-656). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction