missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Kindlin 3 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA566753

Product Code. 15999434

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MADSSYTSEVQ AILAFLSLQR TGSGGPGNHP HGPDASAEGL NPYGLVAPRF QRKFKAKQLT Highest antigen sequence identity to the following orthologs - mouse 84%, rat 84%.

The three KINDLINs are a novel family of focal adhesion proteins, localizing to integrin adhesion sites. The KINDLIN proteins are composed of a centrally located FERM domain interrupted by a pleckstrin homology (PH) domain. KINDLIN1 and KINDLIN2 have been shown to play an essential role in integrin-mediated adhesion and spreading. In contrast to the widely expressed KINDLIN1 and KINDLIN2, KINDLIN3 is restricted to hematopoietic cells and is particularly abundant in megakaryocytes and platelets. Several reports describe a transcriptional misregulation of KINDLINs in various types of cancer. A recent study demonstrates that KINDLIN3 is essential for platelet integrin activation and subsequent integrin outside-in signaling, suggesting it may serve as a potential target for the design of therapeutics aimed at specifically disrupting integrin activation in platelets and leukocytes.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Kindlin 3
Polyclonal
Unconjugated
Fermt3
C79673; fermitin family homolog 3; fermitin family homolog 3 (Drosophila); fermitin family member 3; Fermt3; Kind3; kindlin 3; Kindlin3; Kindlin-3; MGC10966; MIG-2; MIG2B; MIG2-like protein; RGD1310168; UNC-112 related protein 2; UNC112C; unc-112-related protein 2; URP2; URP2SF
Rabbit
Antigen affinity chromatography
RUO
83706
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q86UX7
Fermt3
Recombinant Human Kindlin 3. Recombinant protein control fragment (Product #RP-107411).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.