missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Kindlin 3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12848772
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12848772 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12848772 Supplier Invitrogen™ Supplier No. PA520598

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is rat spleen tissue lysate. PA5-20598 can be used with blocking peptide PEP-0718.

The three KINDLINs are a novel family of focal adhesion proteins, localizing to integrin adhesion sites. The KINDLIN proteins are composed of a centrally located FERM domain interrupted by a pleckstrin homology (PH) domain. KINDLIN1 and KINDLIN2 have been shown to play an essential role in integrin-mediated adhesion and spreading. In contrast to the widely expressed KINDLIN1 and KINDLIN2, KINDLIN3 is restricted to hematopoietic cells and is particularly abundant in megakaryocytes and platelets. Several reports describe a transcriptional misregulation of KINDLINs in various types of cancer. A recent study demonstrates that KINDLIN3 is essential for platelet integrin activation and subsequent integrin outside-in signaling, suggesting it may serve as a potential target for the design of therapeutics aimed at specifically disrupting integrin activation in platelets and leukocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kindlin 3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Fermt3
Gene Accession No. Q86UX7, Q8K1B8
Gene Alias C79673; fermitin family homolog 3; fermitin family homolog 3 (Drosophila); fermitin family member 3; Fermt3; Kind3; kindlin 3; Kindlin3; Kindlin-3; MGC10966; MIG-2; MIG2B; MIG2-like protein; RGD1310168; UNC-112 related protein 2; UNC112C; unc-112-related protein 2; URP2; URP2SF
Gene Symbols Fermt3
Host Species Rabbit
Immunogen A 19 amino acid peptide near the carboxy terminus of the human KINDLIN3.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108101, 309186, 83706
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.