Learn More
Kindlin 2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA546156
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: TSISCYKSKE ESSGTPAHQM NLRGCEVTPD VNISGQKFNI KLLIPVAEGM Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%
Kindlin 2 Polyclonal antibody specifically detects Kindlin 2 in Human samples. It is validated for Western Blot.Specifications
Kindlin 2 | |
Polyclonal | |
Unconjugated | |
FERMT2 | |
AA960555; EGK_18193; Fermitin family homolog 2; fermitin family homolog 2 (Drosophila); fermitin family member 2; fermt2; im:7145859; im:7145859 protein; KIND2; kindlin 2; kindlin-2; LOC616981 protein; MIG2; mig-2; mitogen inducible 2; mitogen inducible gene 2 protein; mitogen-inducible gene 2 protein; PH domain-containing family C member 1; pleckstrin homology domain containing, family C (with FERM domain) member 1; pleckstrin homology domain containing, family C member 1; pleckstrin homology domain-containing family C member 1; Plekhc1; UNC112; UNC112B | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10979 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96AC1 | |
FERMT2 | |
Synthetic peptide directed towards the C-terminal of human FERMT2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |