Learn More
DWK Life Sciences Kimble™ Closures for Sample Vials
Black phenolic closure with PTFE-lined cap
Brand:  DWK Life Sciences 73802-20400
Code : NEW
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10701574
  • £179.00 / Pack of 144
Estimated Shipment: 19-08-2024
to see stock.
Add to basket
Description
Description
Black phenolic closure with PTFE-lined cap
  • Works with media bottles and screw-thread vials
  • Special black phenolic material (wood filled) very resistant to the effects of temperature and steam encountered in autoclaving
  • Cemented-in PTFE-faced rubber liner is provided
Specifications
Specifications
Screw Cap
PTFE, Rubber
Black
20-400
Phenolic