Learn More
DWK Life Sciences Kimble™ Open-Top PolypropyleneScrew Thread Closures with Bonded White PTFE Liners
Intended for single-use injection
POA
Specifications
Material (Liner) | PTFE |
---|---|
Material | Polypropylene |
Autoclavable | Autoclavable |
Description
- Manufactured using the highest quality raw materials
- Designed for an ideal fit with Kimble Chase autosampler vials
- Pre-assembled caps and septa minimizes contamination due to handling
Specifications
PTFE | |
Polypropylene | |
Autoclavable |