Learn More
DWK Life Sciences Kimble™ Kontes™ Ultra-Ware™ Three-Hole Cap Systems
Product Code. 11893053
Click to view available options
Thread Size (Metric):
GL 45
Unit Size:
Each
Product Code. 11893053
Brand: DWK Life Sciences 9539135000
Only null left
Add to basket
Add to basket
Borosilicate glass and PTFE construction give excellent chemical resistance to HPLC mobile phases
  • Supplied with three-hole cap, conical bottom reservoir, standard fittings kit, and PTFE tubing
  • Reservoir has autoclavable plastic safety coating that absorbs UV up to 385nm—preserves UV-sensitive mobile-phase components
  • PTFE cap body is supplied with a TFE/propylene O-ring
  • Connecting threads on top and bottom of cap use 1/4-28 flangeless fittings and adapters Filtration of the mobile phase directly into the reservoir requires additional purchase of a filtration cap and appropriate solvent pickup adapter
TRUSTED_SUSTAINABILITY
Specifications
Material PTFE
No. per Case 1 Cs.
Thread Size (Metric) GL 45
Neck Finish 53 mm
Product Type Multiple Distributor Screw Cap
Autoclavable Autoclavable