missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Kontes™ Micro-Scale Tissue Grinder, All Glass

Product Code. 10161532
Change view
Click to view available options
Capacity:
0.006 oz., 0.2mL
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity unitSize
10161532 0.006 oz., 0.2mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10161532 Supplier DWK Life Sciences Supplier No. 8854700000
  • ONLINE EXCLUSIVE PRICE
    £412.65
    £360.85 / Pack of 6
    Save £51.80
    12% Off
View all promotions
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy

With ground glass pestle

 

  • Manufactured from borosilicate glass 3.3
  • Designed for dispersing small quantities of tissue in buffer solution
  • Working capacity is 0.2 mL 
  • All-glass tube and pestle
  • PTFE bearing prevents misalignment and avoids aerosol formation

 

TRUSTED_SUSTAINABILITY

Specifications

Diameter (English) Shaft 0.11 in.
Diameter (Metric) Shaft 3mm
Capacity (English) 0.006 oz.
Capacity (Metric) 0.2 mL
Material Ground Glass
Product Type Tissue Grinder
For Use With (Application) Designed for dispersing small quantities of tissue in Buffer solution
Length (English) Pestle 2.9 in.
Length (Metric) Pestle 75mm
Length (English) Tube 1.2 in.
Length (Metric) Tube 33mm
Tube Diameter (English) Outside 0.31 in.
Tube Diameter (Metric) Outside 8mm
Capacity 0.006 oz., 0.2mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.