missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences™ Kimble™ Kontes™ MIDI-VAP 4000 Component Replacement Parts

Product Code. 16577279
Change view
Click to view available options
Type:
Reaction/Absorption Tube
For Use With (Equipment):
MIDI-VAP 4000 Cyanide System
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type For Use With (Equipment) Unit Size
16577279 Reaction/Absorption Tube MIDI-VAP 4000 Cyanide System Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16577279 Supplier DWK Life Sciences™ Supplier No. 4794550050
  • ONLINE EXCLUSIVE PRICE
    £87.15
    £79.04 / Pack of 1
    Save £8.11
    9% Off
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Component parts for both ammonia and phenol distillation system

Component parts for both ammonia and phenol distillation system

  • For use with MIDI-VAP 4000 Distillation Apparatus

Specifications

Includes All fittings
For Use With (Application) Distillation systems
Type Reaction/Absorption Tube
Capacity 1.6 oz., 50mL
Capacity (Metric) 50mL
For Use With (Equipment) MIDI-VAP 4000 Cyanide System
Standard Taper Joint 29/42
Capacity (English) 1.6 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.