Learn More
DWK Life Sciences Kimble™ Kontes™ Chromatography Column ETFE Nut for 1/8 in. Tubing
Click to view available options
Material:
PTFE
Description
- ETFE Flangeless Nut
- 1/4-28 Thread, for 1/8 in. and 1.8mm OD Tubing
- Use with the 420822-Series Flangeless Ferrules
- Case of 10
- Replacement Parts in ULTRA-WARE™ Standard Fittings Kit
Specifications
Type | Nut |
For Use With (Equipment) | HPLC cap system |
Material | PTFE |
Quantity | 1 Cs. |