Learn More
DWK Life Sciences Kimble™ Black Phenolic Screw-Thread Closures
Highly resistant to temperature and steam during autoclaving
Brand: DWK Life Sciences 73802-13415
Code : 40
Additional Details : Weight : 0.80000kg
Description
- Cemented-in PTFE-faced 14B white rubber liner
- Three screw-thread finishes
Specifications
Screw Cap | |
Bottle | |
Phenolic Resin | |
13-415 | |
Autoclavable |
PTFE/14B Rubber | |
Black | |
500 Cs. | |
ASTM E982 |