missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Black Phenolic Screw Caps

Product Code. 17721078
Change view
Click to view available options
Neck Finish:
15-415
38-430
Certifications/Compliance:
ASTM E982, Class A
ASTM E982, Class B
No. per Case:
150 Cs.
300 Cs.
Unit Size:
Pack of 150
Pack of 300
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Neck Finish Certifications/Compliance No. per Case unitSize
17721078 38-430 ASTM E982, Class A 150 Cs. Pack of 150
12802033 15-415 ASTM E982, Class B 300 Cs. Pack of 300
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17721078 Supplier DWK Life Sciences Supplier No. 45066B38
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Bond between liner and cap retains adhesive properties even after repeated autoclaving

With PTFE-faced rubber liner

  • Identified in ASTM™ specification E 982-94, Class B requirements
  • With 14B rubber liner (SBR)
    • Has fewer threads than regular GPI caps for easier application and removal
    • Not recommended for use with standard GPI finishes usually found on blown-molded bottles
    • Identified in ASTM specification E 982-94, Class A requirements

    TRUSTED_SUSTAINABILITY

    Specifications

    Color Black
    Height (Metric) With Closure 26mm
    Material Phenolic Resin
    No. per Case 150 Cs.
    Neck Finish 38-430
    Certifications/Compliance ASTM E982, Class A
    For Use With (Equipment) Bottle
    Closure Liner 14B Rubber
    Autoclavable Autoclavable
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.