missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark Professional™ Kimtech™ A5 Sterile Hood
GREENER_CHOICE

Sterile cleanroom hoods feature innovative CLEAN-DON™ technology for simpler aseptic donning. Kimtech™ A5 Sterile Hood suitable for EU GMP ISO 5 Grade A sterile cleanrooms and individually vacuum-packed.

£283.00

Specifications

Product Type Open Face
Size Universal
Material Polypropylene
Color White
Disposable Single-use
View More Specs

Products 1
Product Code Brand Closure Type Price Quantity & Availability  
Product Code Brand Closure Type Price Quantity & Availability  
11358041
GREENER_CHOICE
View Documents Promotion Details
Kimberly-Clark Professional™
25797
None
£283.00
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment: 25-10-2024
to see stock.
Add to basket
 
Description

Description

  • Suitable for use in EU GMP ISO 5 Class A sterile cleanroom environments and laboratories
  • Advanced CLEAN-DON™ technology for fast, simple and compliant aseptic donning that combines two donning steps into one
  • Comfortable hoods are provided in a universal size
  • Made from low-lint breathable SMS fabric with a cloth-like feel
  • Contamination protection is ensured through high quality sterility assured material with high-strength bound and triple-stitched seams, and secure ties
  • Individually vacuum-packed and efficiently double-bagged to save space
  • PPE Cat III according to Regulation (EU) 2016/425
  • Type 6 (PB) certified
  • Certificate of Conformance by lot available online
  • Certificate of Sterility by lot available online

Order Info

Gamma sterilized and individually bagged. Certificate of Irradiation available.

Specifications

Specifications

Open Face
Polypropylene
Single-use
Sterile
100/Cs.
Universal
White
Yes
Triple-bagged, Vacuum Packaged
Class 100 (ISO 5)
Videos
SDS
Documents

Documents

Product Certifications