missing translation for 'onlineSavingsMsg'
Learn More

KleenGuard™ G40 Foam Nitrile Coated Multi-Purpose Gloves

Product Code. 11720674
Change view
Click to view available options
Size:
2X-Large
Unit Size:
12 pairs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
11720674 2X-Large 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11720674 Supplier KleenGuard™ Supplier No. 40229
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

KleenGuard™ G40 Foam Nitrile Coated Multi-Purpose Gloves are designed for jobs that require strength, grip, and dexterity. Seamless knit nylon shell is breathable for comfort, with a foam nitrile coating for enhanced grip capabilities.

  • Designed for jobs where strength, grip and dexterity are a must
  • Protective gloves feature oil and industrial grease grip for general purpose tasks
  • Maintain a high degree of abrasion resistance (EN 388) even after 5 wash cycles
  • 25% better productivity than leather gloves with similar capabilities
  • Seamless knit nylon on the back of the hand is breathable for a comfortable fit
  • Silicone-free
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EN 388
Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Nitrile Foam
Color Black
Exterior Finish Partially Textured
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size 2X-Large
Style Full-hand Mitt
For Use With (Application) Automotive Assembly, Machine Repairs, and Parts Handling
Glove Cuff Style Knit
Series G40
Length (English) 10.2 in.
Length (Metric) 26 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.