missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark Professional™ KleenGuard™ G80 Nitrile Chemical Resistant Gloves

Product Code. 12621496
Click to view available options
:
Size 11
This item is not returnable. View return policy

Product Code. 12621496

Brand: Kimberly-Clark Professional™ 94449

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Green, hand-specific gloves provide maximum protection against chemical and mechanical hazards. G80 Chemical Resistant Latex-Free Gloves include improved, more consistent cotton flock lining for added comfort.

  • Hand specific latex-free gloves protect hands against chemical or mechanical hazards
  • Diamond embossed fingertips offer enhanced wet and dry grip
  • Flock-lined for easy donning and extra comfort in extended use
  • Improved AQL of 0.65 versus 1.5, resulting in fewer pinholes
  • Modified Nitrile formulation for more durability and enhanced chemical resistance.
  • Now available in 5 bulk-packed poly bags versus individually wrapped pairs
  • Ideal for: providing PPE category 3 protection, level 4 abrasion resistance and durability in manufacturing, transport, construction, agriculture cleaning and janitorial work, when handling chemicals, oils, grease, acids, caustics or solvents

Specifications

Certifications/Compliance FDA, ASTM D3767, ASTM D412, ASTM D5151, ASTM D6319
Certifications/Compliance (Europe) CE 0120
Chlorinated No
Color Green
Latex Free Yes
Material Nitrile
Size 2X-Large
Thickness (English) 15 mil
Thickness (Metric) 0.38 mm
EN388 Standard EN 388:4101X
EN 421 No
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES JKLNOP
EN ISO 374 - 5 Bacteria, Fungi only
For Use With (Application) Acids, bases, Solvents and Caustics
Grip Design Rough
Liner Material Cotton Flock
Lining Lined
Model 94449
Series KleenGuard™ G80
Length (English) 13 in.
Length (Metric) 33 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.