Learn More
DiscoverX KILR™ Retroparticles for Suspension Cells (G418)
Product Code. 17384829
Encompass
Change view
Click to view available options
Quantity:
4 Vials
Unit Size:
4 ampoules
Product Code. Quantity Unit Size
17384829 4 Vials 4 ampoules
1 options
Product Code. 17384829 Supplier DiscoverX Supplier No. 970002
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The KILR™ Retroparticles for Suspension Cells (G418) are retroparticles for transducing cells of interest with PL to allow for cytotoxicity assays. Contains 4 vials of cryopreserved retroparticles. Resists G418.
The KILR™ Retroparticles for Suspension Cells (G418) are MMLV particles designed to transduce suspension cells. This allows production of PL-tagged, G418-resistant target cells of interest for use in cytotoxicity assays involving detection of cell lysis. This product uses EFC technology. The retroparticle mix can be used to transduce target cells with a PL-tagged housekeeping gene. Target cell death in a cytotoxicity assay will release PL to the media, where EA contained in the detection reagent will produce EFC signal. These retroparticles ship as 4 vials of cryopreserved retroparticles.
Specifications
For Use With (Application) Retroparticles
Product Type Retroparticles
Quantity 4 Vials