missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX KILR™ Jurkat Cell Pool
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
Description
The KILR Jurkat cell pool is a stable, engineered target cell pool which more closely reflects the heterogeneous native protein expression of tumor environments, used to measure direct cell death mediated by immunooncology treatments (e.g. checkpoint inhibitors, checkpoint agonists, bispecifics, CAR-T, and more) via Antibody Dependent Cell-mediated Cytotoxicity (ADCC), Complement Dependent Cytotoxicity (CDC), and Antibody Dependent Cell Phagocytosis (ADCP). This product uses EFC technology. The cell pool is a heterogeneous population of Jurkat cells that overexpress a PL-tagged housekeeping gene. Once the cells have been lysed by effector cells, PL is released to the media, where EA contained in the detection kit produces an EFC signal proportional to the level of PL released. This allows for calculation of percent lysis of the KILR cell pool. This stable cell pool is provided as two vials of cryopreserved cells.
Specifications
Specifications
| Assay Entry | Cytoxicity |
| Cell Line | Jurkat |
| For Use With (Application) | Cell Pools |
| Quantity | 2 Vials |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction