missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kilon/NEGR1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-32529
This item is not returnable.
View return policy
Description
Kilon/NEGR1 Polyclonal specifically detects Kilon/NEGR1 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Kilon/NEGR1 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| NEGR1 | |
| Recombinant protein encompassing a sequence within the center region of human NEGR1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 257194 | |
| Mouse | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.3mg/mL | |
| Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| a kindred of IgLON, IgLON family member 4, IGLON4DMML2433, KILONMGC46680, neuronal growth regulator 1, neurotractin, Ntra | |
| Rabbit | |
| 39 kDa | |
| 0.1 mL | |
| Primary | |
| Tested in Mouse. Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction