missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIFC3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10125-2-AP-20UL
This item is not returnable.
View return policy
Description
KIFC3 belongs to the large superfamily of kinesins, molecular motors that use the energy of ATP hydrolysis to translocate cargoes along microtubules. Members share extensive homology within a globular domain containing the microtubule- and ATP-binding sites and have a coiled-coil stalk domain that mediates oligomerization. Different kinesin family members participate in specific and diverse motile processes, such as cell division, organelle transport, and nuclear movement (Hoang et al., 1998).Specifications
| KIFC3 | |
| Polyclonal | |
| Unconjugated | |
| KIFC3 | |
| KIFC3, kinesin family member C3, Kinesin like protein KIFC3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16582, 307644, 3801 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O35231, Q9BVG8 | |
| KIFC3 | |
| KIFC3 Fusion Protein Ag0172 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction