missing translation for 'onlineSavingsMsg'
Learn More

KIFC1 Rabbit anti-Human, Clone: S03-3E4, Novus Biologicals™

Product Code. 18378574 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18378574 100 μg 100µL
18303726 25 μg 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18378574 Supplier Novus Biologicals Supplier No. NBP315052100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

KIFC1 Monoclonal antibody specifically detects KIFC1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIFC1
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone S03-3E4
Conjugate Unconjugated
Dilution Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide
Gene Alias HSETMGC1202, kinesin family member C1, kinesin-like 2, Kinesin-like protein 2, kinesin-like protein KIFC1, Kinesin-related protein HSET, KNSL2MGC149736, MGC149737
Host Species Rabbit
Immunogen A synthetic peptide of human KIFC1 (Uniprot#: Q9BW19)
Purification Method Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 3833
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.