missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KIFC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564050
This item is not returnable.
View return policy
Description
Immunogen sequence: EEKERRLQTS EAALSSSQAE VASLRQETVA QAALLTEREE RLHGLEM Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 66%.
Minus end-directed microtubule-dependent motor required for bipolar spindle formation (PubMed:15843429). May contribute to movement of early endocytic vesicles. Regulates cilium formation and structure. [UniProt].
Specifications
| KIFC1 | |
| Polyclonal | |
| Unconjugated | |
| Kifc1 | |
| 100042970; EMBL:AAH83827.1, ECO:0000312; Gm4137; HSET; Kifc1; kifc1 {ECO:0000312; Kifc4; Kifc5a; Kifc5b; kinesin family c-terminal 5A; kinesin family member C1; kinesin motor protein KIFC4; kinesin-like 2; kinesin-like 2a; kinesin-like protein 2; Kinesin-like protein KIFC1; kinesin-related protein 1; Kinesin-related protein HSET; kinesin-related protein KIFC5A; kinesin-related protein KRP1; KNSL2; Knsl2a; Krp1; RGD:1359118}; Tctex7; Tctex-7; Tctex7a; Tctex-7A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3833 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BW19 | |
| Kifc1 | |
| Recombinant protein corresponding to Human KIFC1. Recombinant protein control fragment (Product #RP-103818). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction