missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIFAP3 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97496FR
This item is not returnable.
View return policy
Description
KIFAP3 Polyclonal specifically detects KIFAP3 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| KIFAP3 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| dJ190I16.1, FLJ22818, KAP-3, KIF3AP, kinesin-associated protein 3, small G protein GDP dissociation stimulator, SMAPKAP3Smg-GDS, Smg GDS-associated protein | |
| Produced in rabbits immunized with E. coli-derived Human KIFAP3 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 22920 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction