missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIFAP3 Antibody (0H5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-42988
This item is not returnable.
View return policy
Description
KIFAP3 Monoclonal antibody specifically detects KIFAP3 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
| KIFAP3 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 22920 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |
| ELISA, Immunohistochemistry, Western Blot | |
| 0H5 | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry 1:20 - 1:200 | |
| dJ190I16.1, FLJ22818, KAP-3, KIF3AP, kinesin-associated protein 3, small G protein GDP dissociation stimulator, SMAPKAP3Smg-GDS, Smg GDS-associated protein | |
| kinesin-associated protein 3 - (Uniprot# Q92845) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction