missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
KIF7 Polyclonal antibody specifically detects KIF7 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | KIF7 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 700 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EQYK340, kinesin family member 7, kinesin-like protein KIF7, MGC120653, MGC138476, MGC138478, UNQ340 |
| Host Species | Rabbit |
| Immunogen | The immunogen for this product maps to a region between residue 1225 and 1275 of human kinesin family member 7 using the numbering given in entry NP_940927.2 (GeneID 374654). |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?