missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17290-1-AP-150UL
This item is not returnable.
View return policy
Description
KIF6 belongs to the kinesin-like protein family. There are four named isoforms. The KIF6 719Arg variant has been associated with an increased risk of coronary disease and an increased response to statin therapy.Specifications
| KIF6 | |
| Polyclonal | |
| Unconjugated | |
| KIF6 | |
| C6orf102, KIF6, kinesin family member 6, Kinesin like protein KIF6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 171291, 221458, 319991 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.16 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6ZMV9 | |
| Kif6 | |
| KIF6 Fusion Protein Ag11164 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction