missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF5B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21632-1-AP-150UL
This item is not returnable.
View return policy
Description
Microtubule-dependent motor required for normal distribution of mitochondria and lysosomes. Can induce formation of neurite-like membrane protrusions in non-neuronal cells in a ZFYVE27-dependent manner. Regulates centrosome and nuclear positioning during mitotic entry. During the G2 phase of the cell cycle in a BICD2-dependent manner, antagonizes dynein function and drives the separation of nuclei and centrosomes. Required for anterograde axonal transportation of MAPK8IP3/JIP3 which is essential for MAPK8IP3/JIP3 function in axon elongation.Specifications
| KIF5B | |
| Polyclonal | |
| Unconjugated | |
| KIF5B | |
| KIF5B, Kinesin 1 heavy chain, kinesin family member 5B, KINH, KNS, KNS1, Ubiquitous kinesin heavy chain, UKHC | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 117550, 16573, 3799 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.53 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P33176, Q2PQA9, Q61768 | |
| KIF5B | |
| KIF5B Fusion Protein Ag16250 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction